Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

27579-1G 27579-25G 27579-500MG 27579-5G
76929-570EA 201.58 USD InStock
76929-570 76929-572 76929-574 76929-576
4-Bromo-3,5-dimethylphenylboronic acid pinacol ester ≥97%
4-Bromo-3,5-dimethylphenylboronic acid pinacol ester


Formula: C₁₄H₂₀BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD12405336
CAS Number: 1073338-97-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty