Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formyl-3,5-dimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

28318-1G 28318-500MG 28318-5G
76929-754EA 593.05 USD InStock
76929-754 76929-756 76929-758
4-Formyl-3,5-dimethylbenzoic acid ≥97%
4-Formyl-3,5-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD20644239
CAS Number: 538367-60-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty