Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

28915-25G 28915-5G
76929-920EA 418 USD InStock
76929-920 76929-922
4-Fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%
4-Fluoro-3-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227055
CAS Number: 425378-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty