Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

29278-1G 29278-25G 29278-500MG 29278-5G
76929-984EA 320.4 USD InStock
76929-984 76929-986 76929-988 76929-990
4-Fluoro-3-methoxy-5-methylbenzaldehyde ≥97%
4-Fluoro-3-methoxy-5-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28977309
CAS Number: 1785229-80-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty