Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Carboxy-2,4-dichlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

29293-1G 29293-500MG 29293-5G
76930-000EA 485.9 USD InStock
76930-000 76930-002 76930-004
5-Carboxy-2,4-dichlorophenylboronic acid ≥97%
5-Carboxy-2,4-dichlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BCl₂O₄
MW: 234.829 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806031
CAS Number: 2121514-46-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty