Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-chloropyridine-3-boronic acid pinacol ester ≥95%

Supplier: AOB CHEM USA
Warning

30200-1G 30200-25G 30200-500MG 30200-5G
76930-194EA 379.81 USD InStock
76930-194 76930-196 76930-198 76930-200
5-Bromo-4-chloropyridine-3-boronic acid pinacol ester ≥95%
5-Bromo-4-chloropyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₄BBrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD26518940
CAS Number: 2121514-13-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty