Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-ethoxy-3-pyridineboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

30324-1G 30324-25G 30324-500MG 30324-5G
76930-224EA 334.19 USD InStock
76930-224 76930-226 76930-228 76930-230
5-Chloro-2-ethoxy-3-pyridineboronic acid ≥95%
5-Chloro-2-ethoxy-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₇H₉BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13195643
CAS Number: 1217500-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty