Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methylphenylboronic acid pinaco ester ≥97%

Supplier: AOB CHEM USA
Warning

30401-1G 30401-25G 30401-5G
76930-290EA 92.3 USD InStock
76930-290 76930-292 76930-294
4-Bromo-3-methylphenylboronic acid pinaco ester ≥97%
4-Bromo-3-methylphenylboronic acid pinaco ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD29088160
CAS Number: 2121511-60-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty