Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluoro-2-methylphenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

30483-1G 30483-25G 30483-500MG 30483-5G
76930-332EA 218.55 USD InStock
76930-332 76930-334 76930-336 76930-338
4-Bromo-3-fluoro-2-methylphenol ≥97% research grade
4-Bromo-3-fluoro-2-methylphenol


Formula: C₇H₆BrFO
MW: 205.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414511
CAS Number: 1262516-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty