Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethoxy-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31148-1G 31148-25G 31148-5G
76930-576EA 114.58 USD InStock
76930-576 76930-578 76930-580
5-Ethoxy-2-fluorophenylboronic acid pinacol ester ≥97%
5-Ethoxy-2-fluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD28122695
CAS Number: 2121512-83-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty