Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluoro-2-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31251-1G 31251-25G 31251-500MG 31251-5G
76930-648EA 290.69 USD InStock
76930-648 76930-650 76930-652 76930-654
4-Chloro-3-fluoro-2-methoxyphenylboronic acid pinacol ester ≥97%
4-Chloro-3-fluoro-2-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₇BClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22415058
CAS Number: 1628524-83-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty