Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methoxy-3-methylbenzaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

31801-1G 31801-500MG 31801-5G
76930-744EA 598.35 USD InStock
76930-744 76930-746 76930-748
4-Fluoro-2-methoxy-3-methylbenzaldehyde ≥95%
4-Fluoro-2-methoxy-3-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415111
CAS Number: 1824411-31-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty