Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-2-thiophenecarboxylic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

32756-25G 32756-5G
76931-030EA 453.01 USD InStock
76931-030 76931-032
5-Formyl-2-thiophenecarboxylic acid ≥95% research grade
5-Formyl-2-thiophenecarboxylic acid


Formula: C₆H₄O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00191437
CAS Number: 4565-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty