Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-4'-methoxybiphenyl ≥97% research grade

Supplier: AOB CHEM USA
Warning

32927-1G 32927-25G 32927-5G
76931-066EA 62.6 USD InStock
76931-066 76931-068 76931-070
4-Hydroxy-4'-methoxybiphenyl ≥97% research grade
4-Hydroxy-4'-methoxybiphenyl


Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00191514
CAS Number: 16881-71-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty