Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-(methylthio)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

34924-1G 34924-25G 34924-500MG 34924-5G
76931-554EA 291.75 USD InStock
76931-554 76931-556 76931-558 76931-560
4-Chloro-3-(methylthio)benzoic acid ≥97%
4-Chloro-3-(methylthio)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11501903
CAS Number: 166811-60-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty