Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-3-(meththio)phenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

35419-1G 35419-25G 35419-500MG 35419-5G
76931-668EA 292.81 USD InStock
76931-668 76931-670 76931-672 76931-674
5-Chloro-3-(meththio)phenylboronic acid pinacol ester ≥97%
5-Chloro-3-(meththio)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BClO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD16618973
CAS Number: 1256360-24-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty