Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-isopropoxy-5-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

37291-1G 37291-25G 37291-500MG 37291-5G
76932-006EA 421.18 USD InStock
76932-006 76932-008 76932-010 76932-012
4-Fluoro-3-isopropoxy-5-methylbenzoic acid ≥97%
4-Fluoro-3-isopropoxy-5-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632187
CAS Number: 2404733-89-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty