Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-isobutoxyphenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

39804-25G 39804-5G
76932-334EA 453.01 USD InStock
76932-334 76932-336
4-Chloro-2-isobutoxyphenylboronic acid ≥95%
4-Chloro-2-isobutoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₄BClO₃
MW: 228.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015772
CAS Number: 1256355-06-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty