Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-3,5-difluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

40256-1G 40256-25G 40256-500MG 40256-5G
76932-374EA 153.84 USD InStock
76932-374 76932-376 76932-378 76932-380
4-Ethoxy-3,5-difluorophenylboronic acid ≥97%
4-Ethoxy-3,5-difluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₉BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD20231440
CAS Number: 2096333-93-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty