Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-6-methoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

45646-1G 45646-25G 45646-500MG 45646-5G
76932-498EA 245.07 USD InStock
76932-498 76932-500 76932-502 76932-504
4-Bromo-2-fluoro-6-methoxybenzoic acid ≥97%
4-Bromo-2-fluoro-6-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆BrFO₃
MW: 249.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22570393
CAS Number: 1472104-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty