Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromofuran-3-carbaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

46641-1G 46641-25G 46641-500MG 46641-5G
76932-656EA 368.14 USD InStock
76932-656 76932-658 76932-660 76932-662
4-Bromofuran-3-carbaldehyde ≥97%
4-Bromofuran-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318958
CAS Number: 164513-46-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty