Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(thiophen-3-yl)-3-(trifluoromethyl)-1H-pyrazole ≥97%

Supplier: AOB CHEM USA
Warning

46794-1G 46794-25G 46794-500MG 46794-5G
76932-682EA 409.51 USD InStock
76932-682 76932-684 76932-686 76932-688
5-(thiophen-3-yl)-3-(trifluoromethyl)-1H-pyrazole ≥97%
5-(thiophen-3-yl)-3-(trifluoromethyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₈H₅F₃N₂S
Storage Temperature: Ambient
MDL Number: MFCD07779628
CAS Number: 2408886-65-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty