Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-ethoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

49698-1G 49698-25G 49698-500MG 49698-5G
76932-944EA 215.37 USD InStock
76932-944 76932-946 76932-948 76932-950
4-Chloro-2-ethoxybenzaldehyde ≥97%
4-Chloro-2-ethoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24676068
CAS Number: 869088-32-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty