Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-2,3-dimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

51449-1G 51449-25G 51449-500MG 51449-5G
76933-038EA 322.52 USD InStock
76933-038 76933-040 76933-042 76933-044
4-Hydroxy-2,3-dimethylbenzoic acid ≥97%
4-Hydroxy-2,3-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20640161
CAS Number: 6021-31-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty