Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-2-methoxybenzonitrile ≥97%

Supplier: AOB CHEM USA
Warning

53336-1G 53336-25G 53336-500MG 53336-5G
76933-276EA 296 USD InStock
76933-276 76933-278 76933-280 76933-282
5-Formyl-2-methoxybenzonitrile ≥97%
5-Formyl-2-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD18394185
CAS Number: 21962-50-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty