Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxy-4-methylbenzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

54470-1G 54470-25G 54470-5G
76933-398EA 81.69 USD InStock
76933-398 76933-400 76933-402
5-Bromo-2-methoxy-4-methylbenzoic acid ≥95% research grade
5-Bromo-2-methoxy-4-methylbenzoic acid


Formula: C₉H₉BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD21234732
CAS Number: 90326-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty