Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(tert-Butyl)-2-ethoxybenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

60866-1G 60866-25G 60866-500MG 60866-5G
76935-112EA 142.17 USD InStock
76935-112 76935-114 76935-116 76935-118
5-(tert-Butyl)-2-ethoxybenzaldehyde ≥97% research grade
5-(tert-Butyl)-2-ethoxybenzaldehyde


Formula: C₁₃H₁₈O₂
MW: 206.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02380389
CAS Number: 681443-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty