Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Chloro-2-methoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOB CHEM USA
Warning

61131-1G 61131-25G 61131-500MG 61131-5G
76935-230EA 434.97 USD InStock
76935-230 76935-232 76935-234 76935-236
5-(4-Chloro-2-methoxyphenyl)thiazol-2-amine ≥95%
5-(4-Chloro-2-methoxyphenyl)thiazol-2-amine
Precaución: for Research Use Only


Formula: C₁₀H₉ClN₂OS
Storage Temperature: Ambient
CAS Number: 2755717-12-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty