Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-3-fluoro-2-methoxyisonicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

64748-1G 64748-25G 64748-500MG 64748-5G
76935-644EA 266.29 USD InStock
76935-644 76935-646 76935-648 76935-650
5-Chloro-3-fluoro-2-methoxyisonicotinic acid ≥97%
5-Chloro-3-fluoro-2-methoxyisonicotinic acid
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₃
MW: 205.5709 g/mol
Storage Temperature: Ambient
CAS Number: 2374718-75-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty