Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-formyl-2-methylbenzonitrile ≥97%

Supplier: AOB CHEM USA
Warning

80987-1G 80987-500MG 80987-5G 80987-25G
76936-018EA 1048.17 USD InStock
76936-018 76936-020 76936-022 77636-216
5-Chloro-4-formyl-2-methylbenzonitrile ≥97%
5-Chloro-4-formyl-2-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₆ClNO
MW: 179.603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34816592
CAS Number: 2386527-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty