Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-dimethoxyisonicotinaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

95106-1G 95106-500MG 95106-5G
76936-112EA 684.29 USD InStock
76936-112 76936-114 76936-116
5-Chloro-2,3-dimethoxyisonicotinaldehyde ≥97%
5-Chloro-2,3-dimethoxyisonicotinaldehyde


Formula: C₈H₈ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20487003
CAS Number: 1305324-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty