Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl thieno[3,2-b]thiophene-2-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

104040-25G 104040-5G
76936-380EA 542.12 USD InStock
76936-380 76936-382
Methyl thieno[3,2-b]thiophene-2-carboxylate ≥97%
Methyl thieno[3,2-b]thiophene-2-carboxylate


Formula: C₈H₆O₂S₂
Storage Temperature: Ambient
MDL Number: MFCD01934498
CAS Number: 98800-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty