Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-chloro-4-(difluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

13440-1G 13440-25G 13440-500MG 13440-5G
76936-732EA 374.5 USD InStock
76936-732 76936-734 76936-736 76936-738
Ethyl 3-chloro-4-(difluoromethoxy)benzoate ≥97%
Ethyl 3-chloro-4-(difluoromethoxy)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₉ClF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD24108569
CAS Number: 1510018-84-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty