Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl 3-chloro-4-(difluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

13450-1G 13450-500MG 13450-5G
76936-748EA 683.22 USD InStock
76936-748 76936-750 76936-752
Benzyl 3-chloro-4-(difluoromethoxy)benzoate ≥97%
Benzyl 3-chloro-4-(difluoromethoxy)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₁ClF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32631765
CAS Number: 2364584-74-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty