Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,2-difluoro-2-(thiophen-2-yl)acetate ≥97%

Supplier: AOB CHEM USA
Warning

147873-1G 147873-500MG 147873-5G
76936-920EA 744.76 USD InStock
76936-920 76936-922 76936-924
Ethyl 2,2-difluoro-2-(thiophen-2-yl)acetate ≥97%
Ethyl 2,2-difluoro-2-(thiophen-2-yl)acetate
Precaución: for Research Use Only


Formula: C₈H₈F₂O₂S
Storage Temperature: Refrigerator
CAS Number: 294182-00-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty