Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

14977-1G 14977-500MG 14977-5G
76936-928EA 500.75 USD InStock
76936-928 76936-930 76936-932
6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid ≥95% research grade
6-Bromo-2-fluoro-3-(methoxycarbonyl)benzeneboronic acid


Formula: C₈H₇BBrFO₄
Storage Temperature: Ambient
MDL Number: MFCD09800873
CAS Number: 957120-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty