Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Isopropoxynicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

18829-1G 18829-25G 18829-5G
76937-386EA 102.91 USD InStock
76937-386 76937-388 76937-390
6-Isopropoxynicotinic acid ≥97% research grade
6-Isopropoxynicotinic acid


Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09811743
CAS Number: 223127-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty