Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-5-fluoroisonicotinate ≥97%

Supplier: AOB CHEM USA
Warning

20254-1G 20254-25G 20254-500MG 20254-5G
76937-614EA 260.99 USD InStock
76937-614 76937-616 76937-618 76937-620
Methyl 3-bromo-5-fluoroisonicotinate ≥97%
Methyl 3-bromo-5-fluoroisonicotinate
Precaución: for Research Use Only


Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698118
CAS Number: 1214325-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty