Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,5-dibromo-4-pyridinecarboxylate ≥97%

Supplier: AOB CHEM USA
Warning

20350-1G 20350-25G 20350-5G
76937-628EA 79.57 USD InStock
76937-628 76937-630 76937-632
Ethyl 2,5-dibromo-4-pyridinecarboxylate ≥97%
Ethyl 2,5-dibromo-4-pyridinecarboxylate
Precaución: for Research Use Only


Formula: C₈H₇Br₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698214
CAS Number: 1214361-41-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty