Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-chloro-2-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

21572-1G 21572-25G 21572-500MG 21572-5G
76937-928EA 281.14 USD InStock
76937-928 76937-930 76937-932 76937-934
Ethyl 4-chloro-2-(trifluoromethyl)benzoate ≥97%
Ethyl 4-chloro-2-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₈ClF₃O₂
MW: 252.618 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06205101
CAS Number: 773139-28-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty