Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-5-methyl-1H-indazole ≥97%

Supplier: AOB CHEM USA
Warning

26188-1G 26188-25G 26188-500MG 26188-5G
76938-814EA 133.68 USD InStock
76938-814 76938-816 76938-818 76938-820
7-Bromo-5-methyl-1H-indazole ≥97%
7-Bromo-5-methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD05664004
CAS Number: 885272-97-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty