Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-2-methylnicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

26501-25G 26501-5G
76938-900EA 424.36 USD InStock
76938-900 76938-902
6-Fluoro-2-methylnicotinic acid ≥97% research grade
6-Fluoro-2-methylnicotinic acid


Formula: C₇H₆FNO₂
MW: 155.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662400
CAS Number: 884494-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty