Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2-ethylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

26688-1G 26688-25G 26688-500MG 26688-5G
76938-950EA 260.99 USD InStock
76938-950 76938-952 76938-954 76938-956
Methyl 4-bromo-2-ethylbenzoate ≥97%
Methyl 4-bromo-2-ethylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490667
CAS Number: 194487-79-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty