Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methyl-1H-indazole-5-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

26759-1G 26759-25G 26759-500MG 26759-5G
76938-966EA 320.4 USD InStock
76938-966 76938-968 76938-970 76938-972
7-Methyl-1H-indazole-5-boronic acid pinacol ester ≥97%
7-Methyl-1H-indazole-5-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27956924
CAS Number: 2121513-33-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty