Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-dichloro-4-iodobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

27660-1G 27660-500MG 27660-5G
76939-184EA 563.34 USD InStock
76939-184 76939-186 76939-188
Methyl 2,6-dichloro-4-iodobenzoate ≥97%
Methyl 2,6-dichloro-4-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₅Cl₂IO₂
MW: 330.935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194679
CAS Number: 1098619-73-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty