Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-2-methoxyisonicotinate ≥97%

Supplier: AOB CHEM USA
Warning

29229-1G 29229-25G 29229-5G
76939-424EA 96.55 USD InStock
76939-424 76939-426 76939-428
Methyl 5-bromo-2-methoxyisonicotinate ≥97%
Methyl 5-bromo-2-methoxyisonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375112
CAS Number: 886365-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty