Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-methyl-2-(propan-2-yloxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

29660-1G 29660-25G 29660-500MG 29660-5G
76939-494EA 215.37 USD InStock
76939-494 76939-496 76939-498 76939-500
Methyl 5-methyl-2-(propan-2-yloxy)benzoate ≥97%
Methyl 5-methyl-2-(propan-2-yloxy)benzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD23115023
CAS Number: 1368384-39-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty