Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methoxy-5-(trifluoromethyl)benzoate ≥95%

Supplier: AOB CHEM USA
Warning

29781-1G 29781-25G 29781-500MG 29781-5G
76939-518EA 221.73 USD InStock
76939-518 76939-520 76939-522 76939-524
Methyl 2-methoxy-5-(trifluoromethyl)benzoate ≥95%
Methyl 2-methoxy-5-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06204233
CAS Number: 177174-47-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty