Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Phenylpyridin-3-yl)boronic acid, pinacol ester ≥95%

Supplier: AOB CHEM USA
Warning

30149-1G 30149-25G 30149-500MG 30149-5G
76939-610EA 178.24 USD InStock
76939-610 76939-612 76939-614 76939-616
(5-Phenylpyridin-3-yl)boronic acid, pinacol ester ≥95%
(5-Phenylpyridin-3-yl)boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₇H₂₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13182146
CAS Number: 1171891-07-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty