Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenoxypyridine-3-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

31413-1G 31413-25G 31413-500MG 31413-5G
76939-932EA 270.53 USD InStock
76939-932 76939-934 76939-936 76939-938
5-Phenoxypyridine-3-boronic acid ≥97%
5-Phenoxypyridine-3-boronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09907982
CAS Number: 850991-33-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty